missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon 15 mL ViewPoint™ Centrifuge Thermochromic Tubes
Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
30.30 € - 53.90 €
Specifications
Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity |
---|---|
Closure Color | Purple |
Color | Clear to Blue color change at 0°C to 4°C |
Capacity (Metric) | 15 mL |
Disposable | Single-use |
Product Code | Brand | No. per Pack | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Pack | Sterility | Price | Quantity & Availability | ||||
16609842
|
Labcon
3210-320-006-9 |
10/IntegraPack™ Bag | Non-sterile |
49.30 €
Pack of 50 |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||||
16619842
|
Labcon
3210-325-006-9 |
10/IntegraPack™ Bag | Sterile |
53.90 €
Pack of 50 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
16629842
|
Labcon
3210-335-006-9 |
25/reusable polypropylene rack | Sterile |
30.30 €
Pack of 50 |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
Description
- Real-time temperature signaling
- Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
- With attached flat screw caps
- Black graduations and white writing surface
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
- Polypropylene USP Class VI resin
- Centrifuging to 12,500 x G
- Freezable to -80°C
- Made in USA
Specifications
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity | |
Clear to Blue color change at 0°C to 4°C | |
Single-use | |
Room Temperature | |
118 mm | |
Conical | |
17 mm | |
Conical Centrifuge Tube |
Purple | |
15 mL | |
ViewPoint™ | |
Autoclavable | |
17 mm | |
Yes | |
2 to 15 in 0.5 increments | |
12,500 x g |