missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble Chase™ Microlink Closures
Click to view available options
Neck Finish:
18-415
Description
- PTFE
- Screw Closure
- For Kimble mixing-bulb Class A volumetric flasks

Specifications
Specifications
Color | Black |
Material | Polypropylene |
No. per Case | 288 Cs. |
Neck Finish | 18-415 |
For Use With (Equipment) | Bottle |
Closure Liner | PTFE/Silicone |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble Chase™ Microlink Closures >
Spot an opportunity for improvement?Share a Content Correction