missing translation for 'onlineSavingsMsg'
Learn More
18-β-Glycyrrhetinic Acid, 98+%, Thermo Scientific Chemicals Product Code.: 10570931

18-β-Glycyrrhetinic Acid, 98+%, Thermo Scientific Chemicals

Product Code. 10570931
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10570931

Brand: Thermo Scientific Acros 120150050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-53-4
Molecular Formula C30H46O4
Molecular Weight (g/mol) 470.69
MDL Number MFCD00003706,MFCD00066716
InChI Key MPDGHEJMBKOTSU-YKLVYJNSSA-N
Synonym enoxolone, glycyrrhetinic acid, uralenic acid, glycyrrhetic acid, 18-beta-glycyrrhetinic acid, glycyrrhetin, arthrodont, jintan, 18beta-glycyrrhetinic acid, rhetinic acidShow MoreShow Less
PubChem CID 10114
ChEBI CHEBI:30853
IUPAC Name (2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acidShow MoreShow Less
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 292°C to 295°C
Color White
Loss on Drying 1% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 3644
Specific Rotation Condition +165° (20°C c=1,CHC3,on dry ba)
Specific Rotation + 165
Formula Weight 470.69
Percent Purity 98+%
Related Substances 2% max. (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 18-β-Glycyrrhetinic acid, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467921 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number120150050,120150250
Lot NumberA0457634 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number120150050,120150250
Lot NumberA0428504 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number120150050,120150250
Lot NumberA0397369 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number120150050,120150250
Lot NumberA0410602 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number120150050,120150250
Product Identifier
  • 18-ß-Glycyrrhetinic acid,

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
18-β-Glycyrrhetinic Acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.