missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Perfluoro-1-decanol, 97%, Thermo Scientific™
45.88 € - 197.00 €
Chemical Identifiers
CAS | 678-39-7 |
---|---|
Molecular Formula | C10H5F17O |
Molecular Weight (g/mol) | 464.122 |
MDL Number | MFCD00039544 |
InChI Key | JJUBFBTUBACDHW-UHFFFAOYSA-N |
Synonym | 1h,1h,2h,2h-perfluoro-1-decanol, 1h,1h,2h,2h-perfluorodecanol, 2-perfluoro-n-octyl ethanol, 1,1,2,2-tetrahydroperfluoro-1-decanol, 2-perfluorooctyl ethanol, 8:2 fluorotelomer alcohol, 1h,1h,2h,2h-perfluorodecan-1-ol, 8:2 ftoh, unii-um7l66ofe1, 1-decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro |
PubChem CID | 69619 |
ChEBI | CHEBI:63919 |
IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol |
SMILES | C(CO)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is a chemical applied to the formation of a barrier and self-healing coating on zinc metal materials. It induces cell death and formation of oxidative or reactive oxygen species in cerebellar granule cells. It is used to make surfactant and polymeric products with a range of commercial applications (may contain small amounts of residual unreacted material). The polymers are primarily used for coating textiles, paper, and carpet for oil, stain, and water repellency.
Solubility
Soluble in chloroform and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
678-39-7 | |
464.122 | |
JJUBFBTUBACDHW-UHFFFAOYSA-N | |
69619 | |
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol |
C10H5F17O | |
MFCD00039544 | |
1h,1h,2h,2h-perfluoro-1-decanol, 1h,1h,2h,2h-perfluorodecanol, 2-perfluoro-n-octyl ethanol, 1,1,2,2-tetrahydroperfluoro-1-decanol, 2-perfluorooctyl ethanol, 8:2 fluorotelomer alcohol, 1h,1h,2h,2h-perfluorodecan-1-ol, 8:2 ftoh, unii-um7l66ofe1, 1-decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro | |
CHEBI:63919 | |
C(CO)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Specifications
678-39-7 | |
112°C to 114°C (10 mmHg) | |
MFCD00039544 | |
1h,1h,2h,2h-perfluoro-1-decanol, 1h,1h,2h,2h-perfluorodecanol, 2-perfluoro-n-octyl ethanol, 1,1,2,2-tetrahydroperfluoro-1-decanol, 2-perfluorooctyl ethanol, 8:2 fluorotelomer alcohol, 1h,1h,2h,2h-perfluorodecan-1-ol, 8:2 ftoh, unii-um7l66ofe1, 1-decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro | |
JJUBFBTUBACDHW-UHFFFAOYSA-N | |
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol | |
69619 | |
464.12 | |
1H,1H,2H,2H-Perfluoro-1-decanol |
45°C to 50°C | |
C10H5F17O | |
2227487 | |
Soluble in chloroform and methanol. | |
C(CO)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
464.122 | |
CHEBI:63919 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 211-648-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures