missing translation for 'onlineSavingsMsg'
Learn More

Econix™ Bio-bins™ Plain Paper Based Non-Sharps Waste Containers for General Use Only
GREENER_CHOICE

Use Econix™ Bio-bin™ General/Non Infectious Bio-bin™ to dispose of single use implements and reduce over filling issues for your non-sharp clinical waste. For use with general waste.

Brand:  Econix™ BEXBBLOOP

 View more versions of this product

Product Code. 13530870

  • 234.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • 96% paper based non-plastic single-use non-sharps waste container
  • Comes fold-flat for easy storage
  • Rigid, leakproof construction
  • Leakproof, built-in lid
  • Built-in absorbent Mat
  • UN 3291 approved
Includes temporary seal Autoclavable
Specifications

Specifications

2 L, 2.5 L
Temporary Seal
White, Green
180 mm, 120 mm
Non-sterile
230 mm
For General Waste
Non-Sharps Waste Container
Not required
Space efficient fold-flat
230 x 110 x 180 mm
230 mm
100 Pk.
Autoclavable
Cardboard, Recycled Paper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.