missing translation for 'onlineSavingsMsg'
Learn More
2,1,3-Benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™ Product Code.: 10101162

2,1,3-Benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10101162
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10101162

Brand: Thermo Scientific Maybridge CC09603DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 114322-14-4
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H3ClN2O3S
Quantity 1 g
Synonym benzofurazan-4-sulfonyl chloride, benzofurazan-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, benzo 1,2,5 oxadiazole-4-sulfonyl chloride, benzo c 1,2,5 oxadiazole-4-sulfonyl chloride, 2,1,3-benzoxadiazole-4-sulphonyl chloride, acmc-20echy, pubchem5461, 4-chlorosulphonylbenzofurazan, benzofurazan4-sulfonyl chloride
InChI Key SIYWVPMYOGMRDW-UHFFFAOYSA-N
SMILES C1=CC2=NON=C2C(=C1)S(=O)(=O)Cl
IUPAC Name 2,1,3-benzoxadiazole-4-sulfonyl chloride
Molecular Weight (g/mol) 218.611
PubChem CID 2735446
Formula Weight 218.62
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole-4-sulfonyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,1,3-Benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.