missing translation for 'onlineSavingsMsg'
Learn More
Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 15480588

Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15480588
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15480588

Brand: Thermo Scientific Alfa Aesar H54275.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114322-14-4
Molecular Formula C6H3ClN2O3S
Molecular Weight (g/mol) 218.611
MDL Number MFCD00068072
InChI Key SIYWVPMYOGMRDW-UHFFFAOYSA-N
Synonym benzofurazan-4-sulfonyl chloride, benzofurazan-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, benzo 1,2,5 oxadiazole-4-sulfonyl chloride, benzo c 1,2,5 oxadiazole-4-sulfonyl chloride, 2,1,3-benzoxadiazole-4-sulphonyl chloride, acmc-20echy, pubchem5461, 4-chlorosulphonylbenzofurazan, benzofurazan4-sulfonyl chlorideShow MoreShow Less
PubChem CID 2735446
IUPAC Name 2,1,3-benzoxadiazole-4-sulfonyl chloride
SMILES C1=CC2=NON=C2C(=C1)S(=O)(=O)Cl

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 218.61
Percent Purity 97%
Chemical Name or Material Benzofurazan-4-sulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA11Y025 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54275.06,H54275.03,H54275.MD
Lot NumberN15H044 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54275.06,H54275.03,H54275.MD
Lot NumberP17A002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54275.06,H54275.03,H54275.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.