missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,1,3-Benzoxadiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 120.11 |
Percent Purity | 97% |
Chemical Name or Material | 2, 1, 3-Benzoxadiazole |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Safety and Handling

Product Identifier
- 2,1,3-Benzoxadiazole
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 2,1,3-Benzoxadiazole
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,1,3-Benzoxadiazole, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction