missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-(1H-Tetrazol-5-yl)phenol, 97%
54.00 € - 494.00 €
Chemical Identifiers
CAS | 51449-77-5 |
---|---|
Molecular Formula | C7H6N4O |
Molecular Weight (g/mol) | 162.152 |
MDL Number | MFCD09037787 |
InChI Key | SQPPXOJPOBGAAU-UHFFFAOYSA-N |
Synonym | phenol,2-2h-tetrazol-5-yl, 2-2h-1,2,3,4-tetrazol-5-yl phenol, 5-2-hydroxyphenyl-1h-tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl phenol, acmc-1akh0, 5-o-hydroxyphenyl tetrazole, 2-1h-tetrazole-5-yl phenol, fpuqyvargfdcgz-uhfffaoysa-n, 5-2-hydroxyphenyl-1h-tetrazol, 2-2h-1,2,3,4-tetraazol-5-yl phenol |
PubChem CID | 42553059 |
IUPAC Name | 6-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,4-dien-1-one |
SMILES | C1=CC(=C2NNN=N2)C(=O)C=C1 |
Description
Chemical Identifiers
51449-77-5 | |
162.152 | |
SQPPXOJPOBGAAU-UHFFFAOYSA-N | |
42553059 | |
C1=CC(=C2NNN=N2)C(=O)C=C1 |
C7H6N4O | |
MFCD09037787 | |
phenol,2-2h-tetrazol-5-yl, 2-2h-1,2,3,4-tetrazol-5-yl phenol, 5-2-hydroxyphenyl-1h-tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl phenol, acmc-1akh0, 5-o-hydroxyphenyl tetrazole, 2-1h-tetrazole-5-yl phenol, fpuqyvargfdcgz-uhfffaoysa-n, 5-2-hydroxyphenyl-1h-tetrazol, 2-2h-1,2,3,4-tetraazol-5-yl phenol | |
6-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,4-dien-1-one |
Specifications
51449-77-5 | |
C7H6N4O | |
phenol,2-2h-tetrazol-5-yl, 2-2h-1,2,3,4-tetrazol-5-yl phenol, 5-2-hydroxyphenyl-1h-tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl phenol, acmc-1akh0, 5-o-hydroxyphenyl tetrazole, 2-1h-tetrazole-5-yl phenol, fpuqyvargfdcgz-uhfffaoysa-n, 5-2-hydroxyphenyl-1h-tetrazol, 2-2h-1,2,3,4-tetraazol-5-yl phenol | |
C1=CC(=C2NNN=N2)C(=O)C=C1 | |
162.152 | |
162.16 | |
2-(1H-Tetrazol-5-yl)phenol |
∽230°C (decomposition) | |
MFCD09037787 | |
SQPPXOJPOBGAAU-UHFFFAOYSA-N | |
6-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,4-dien-1-one | |
42553059 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures