Learn More
2-(2-Bromophenyl)ethylamine, 97%, Thermo Scientific Chemicals
83.60 € - 318.00 €
Chemical Identifiers
CAS | 65185-58-2 |
---|---|
Molecular Formula | C8H10BrN |
Molecular Weight (g/mol) | 200.079 |
MDL Number | MFCD01529874 |
InChI Key | ITRNQMJXZUWZQL-UHFFFAOYSA-N |
Synonym | 2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine |
PubChem CID | 2734091 |
IUPAC Name | 2-(2-bromophenyl)ethanamine |
SMILES | C1=CC=C(C(=C1)CCN)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15407238
|
Thermo Scientific Alfa Aesar
H60739.04 |
2 g |
83.60 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417238
|
Thermo Scientific Alfa Aesar
H60739.09 |
10 g |
318.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
65185-58-2 | |
200.079 | |
ITRNQMJXZUWZQL-UHFFFAOYSA-N | |
2734091 | |
C1=CC=C(C(=C1)CCN)Br |
C8H10BrN | |
MFCD01529874 | |
2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine | |
2-(2-bromophenyl)ethanamine |
Specifications
65185-58-2 | |
253°C | |
1.577 | |
UN2735 | |
2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine | |
C1=CC=C(C(=C1)CCN)Br | |
200.079 | |
200.08 | |
2-(2-Bromophenyl)ethylamine |
1.396 | |
C8H10BrN | |
MFCD01529874 | |
Air Sensitive | |
ITRNQMJXZUWZQL-UHFFFAOYSA-N | |
2-(2-bromophenyl)ethanamine | |
2734091 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.