Learn More
2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals
53.67 € - 1066.31 €
Chemical Identifiers
CAS | 366-18-7 |
---|---|
Molecular Formula | C10H8N2 |
Molecular Weight (g/mol) | 156.19 |
MDL Number | MFCD00006212 |
InChI Key | ROFVEXUMMXZLPA-UHFFFAOYSA-N |
Synonym | 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridylShow More |
PubChem CID | 1474 |
ChEBI | CHEBI:30351 |
IUPAC Name | 2-pyridin-2-ylpyridine |
SMILES | C1=CC=C(N=C1)C1=CC=CC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10080810
|
Thermo Scientific Acros
117500100 |
10 g | Glass bottle |
53.67 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10274570
|
Thermo Scientific Acros
117500250 |
25 g | Glass bottle |
105.06 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530581
|
Thermo Scientific Acros
117501000 |
100 g | Glass bottle |
292.46 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936291
|
Thermo Scientific Acros
117505000 |
500 g | Glass bottle |
1066.31 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
366-18-7 | |
156.19 | |
ROFVEXUMMXZLPA-UHFFFAOYSA-N | |
1474 | |
2-pyridin-2-ylpyridine |
C10H8N2 | |
MFCD00006212 | |
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridylShow More | |
CHEBI:30351 | |
C1=CC=C(N=C1)C1=CC=CC=N1 |
Specifications
366-18-7 | |
100.0 | |
White | |
121°C | |
99% min. (GC) | |
MFCD00006212 | |
16,25 | |
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl | |
ROFVEXUMMXZLPA-UHFFFAOYSA-N | |
2-pyridin-2-ylpyridine | |
1474 | |
1.51 mm2/s (90°C) | |
99+% | |
2, 2'-Dipyridyl |
99.0 | |
69.0°C to 72.0°C | |
272.0°C to 273.0°C | |
Authentic | |
C10H8N2 | |
23,199 | |
15,1241 | |
Solubility in water: slightly soluble (5.5g/L 22°C). Other solubilities: very soluble in ethanol (500g/L 25°C), ether,, benzene, chloroform and petroleum ether | |
C1=CC=C(N=C1)C1=CC=CC=N1 | |
156.19 | |
CHEBI:30351 | |
156.19 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or d
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-674-4
- 2,2``-Dipyridyl
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 2,2-Bipyridine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.