missing translation for 'onlineSavingsMsg'
Learn More
2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals Product Code.: 11936291

2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals

Product Code. 11936291
500 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11936291

Brand: Thermo Scientific Acros 117505000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridylShow MoreShow Less
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2-pyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 69.0°C to 72.0°C
Color White
Boiling Point 272.0°C to 273.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 500 g
UN Number 2811
Beilstein 23,199
Fieser 16,25
Merck Index 15,1241
Solubility Information Solubility in water: slightly soluble (5.5g/L 22°C). Other solubilities: very soluble in ethanol (500g/L 25°C), ether,, benzene, chloroform and petroleum ether
Viscosity 1.51 mm2/s (90°C)
Formula Weight 156.19
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2'-Dipyridyl
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466581 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number117501000,117500100,117500250,117505000
Lot NumberA0465442 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog Number117501000,117500100,117500250,117505000
Lot NumberA0464306 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog Number117501000,117500100,117500250,117505000
Lot NumberA0463675 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number117501000,117500100,117500250,117505000
Lot NumberA0462614 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number117501000,117500100,117500250,117505000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.