missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Thermo Scientific™

82.10 € - 956.00 €

Chemical Identifiers

CAS 165115-15-1
Molecular Formula C6H10N2S
Molecular Weight (g/mol) 142.22
MDL Number MFCD03011601
InChI Key GLEZGSKBHYTSAD-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine
PubChem CID 7536518
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)ethanamine
SMILES CC1=NC(CCN)=CS1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10122844
View Documents
Thermo Scientific Maybridge
CC54613CB
250 mg Amber Glass Bottle
82.10 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10671934
View Documents
Thermo Scientific Maybridge
CC54613DA
1 g Amber Glass Bottle
222.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10737485
View Documents
Thermo Scientific Maybridge
CC54613EA
10 g Amber Glass Bottle
956.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

165115-15-1
142.22
GLEZGSKBHYTSAD-UHFFFAOYSA-N
7536518
CC1=NC(CCN)=CS1
C6H10N2S
MFCD03011601
2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine
2-(2-methyl-1,3-thiazol-4-yl)ethanamine

Specifications

165115-15-1
C6H10N2S
2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine
CC1=NC(CCN)=CS1
142.22
142.23
2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine
97%
MFCD03011601
GLEZGSKBHYTSAD-UHFFFAOYSA-N
2-(2-methyl-1,3-thiazol-4-yl)ethanamine
7536518
97%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.