missing translation for 'onlineSavingsMsg'
Learn More
2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine, 95%, Thermo Scientific Chemicals Product Code.: 15413797

2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine, 95%, Thermo Scientific Chemicals

Product Code. 15413797
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15413797

Brand: Thermo Scientific Alfa Aesar H27843.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5946-39-4
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD03789577
InChI Key MLXBHOCKBUILHN-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepineShow MoreShow Less
PubChem CID 2771762
IUPAC Name 2,3,4,5-tetrahydro-1H-1,4-benzodiazepine
SMILES C1CNC2=CC=CC=C2CN1

Specifications

Melting Point 96°C to 100°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 148.21
Percent Purity 95%
Chemical Name or Material 2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10171110 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog NumberH27843.03,H27843.MD
Lot Number10140114 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27843.03,H27843.MD
Lot Number10132282 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27843.03,H27843.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.