Learn More
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals
40.80 € - 1127.61 €
Chemical Identifiers
CAS | 84-58-2 |
---|---|
Molecular Formula | C8Cl2N2O2 |
Molecular Weight (g/mol) | 227.01 |
InChI Key | HZNVUJQVZSTENZ-UHFFFAOYSA-N |
Synonym | 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile |
PubChem CID | 6775 |
IUPAC Name | 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile |
SMILES | C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10449020
|
Thermo Scientific Acros
113300100 |
10 g | Glass bottle |
40.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366270
|
Thermo Scientific Acros
113301000 |
100 g | Glass bottle |
234.17 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915611
|
Thermo Scientific Acros
113305000 |
500 g | Glass bottle |
1127.61 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
84-58-2 | |
227.01 | |
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile | |
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile |
C8Cl2N2O2 | |
HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
6775 | |
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N |
Specifications
84-58-2 | |
100.0 | |
Yellow to Orange | |
Authentic | |
C8Cl2N2O2 | |
01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120 | |
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile | |
HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile | |
6775 | |
98% | |
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone |
97.5 | |
210.0°C to 217.0°C | |
0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum) | |
97.5% min. (Iodometry) | |
10,902 | |
15,3072 | |
Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride | |
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N | |
227.01 | |
227.01 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective glo
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-542-2
- 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only