Learn More
2,3-Difluorophenol, 98+%, Thermo Scientific Chemicals
30.75 € - 82.61 €
Chemical Identifiers
CAS | 6418-38-8 |
---|---|
Molecular Formula | C6H4F2O |
Molecular Weight (g/mol) | 130.094 |
MDL Number | MFCD00010262 |
InChI Key | RPEPGIOVXBBUMJ-UHFFFAOYSA-N |
Synonym | phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol |
PubChem CID | 80879 |
IUPAC Name | 2,3-difluorophenol |
SMILES | C1=CC(=C(C(=C1)F)F)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458806
|
Thermo Scientific Alfa Aesar
A16971.03 |
1 g |
30.75 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468806
|
Thermo Scientific Alfa Aesar
A16971.06 |
5 g |
82.61 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6418-38-8 | |
130.094 | |
RPEPGIOVXBBUMJ-UHFFFAOYSA-N | |
80879 | |
C1=CC(=C(C(=C1)F)F)O |
C6H4F2O | |
MFCD00010262 | |
phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol | |
2,3-difluorophenol |
Specifications
6418-38-8 | |
53°C to 55°C (25 mmHg) | |
Phenol-like | |
MFCD00010262 | |
2082265 | |
RPEPGIOVXBBUMJ-UHFFFAOYSA-N | |
2,3-difluorophenol | |
80879 | |
≥98% |
34°C to 38°C | |
56°C (132°F) | |
C6H4F2O | |
UN2921 | |
phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol | |
C1=CC(=C(C(=C1)F)F)O | |
130.094 | |
130.09 | |
2,3-Difluorophenol |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.