missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1H-indol-2-ylmethanol, Thermo Scientific™

122.00 € - 725.00 €

Chemical Identifiers

CAS 27640-31-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
InChI Key GRPOFAKYHPAXNP-UHFFFAOYSA-N
Synonym indolin-2-ylmethanol, 2,3-dihydro-1h-indol-2-yl-methanol, 1h-indole-2-methanol, 2,3-dihydro, 2,3-dihydro-1h-indol-2-yl methanol, 2-hydroxymethylindoline, acmc-209gye, 2-hydroxymethyl-indoline, indolin-2-ylmethan-1-ol, rs-2-hydroxymethylindoline, +-2-hydroxymethyl-indoline
PubChem CID 2794664
IUPAC Name 2,3-dihydro-1H-indol-2-ylmethanol
SMILES C1C(NC2=CC=CC=C21)CO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10157042
View Documents
Thermo Scientific Maybridge
MO01098CB
250 mg Amber Glass Bottle
122.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10123822
View Documents
Thermo Scientific Maybridge
MO01098DA
1 g Amber Glass Bottle
298.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10396564
View Documents
Thermo Scientific Maybridge
MO01098DE
5 g Amber Glass Bottle
725.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

27640-31-9
149.193
indolin-2-ylmethanol, 2,3-dihydro-1h-indol-2-yl-methanol, 1h-indole-2-methanol, 2,3-dihydro, 2,3-dihydro-1h-indol-2-yl methanol, 2-hydroxymethylindoline, acmc-209gye, 2-hydroxymethyl-indoline, indolin-2-ylmethan-1-ol, rs-2-hydroxymethylindoline, +-2-hydroxymethyl-indoline
2,3-dihydro-1H-indol-2-ylmethanol
C9H11NO
GRPOFAKYHPAXNP-UHFFFAOYSA-N
2794664
C1C(NC2=CC=CC=C21)CO

Specifications

27640-31-9
C9H11NO
GRPOFAKYHPAXNP-UHFFFAOYSA-N
2,3-dihydro-1H-indol-2-ylmethanol
2794664
97%
97%
indolin-2-ylmethanol, 2,3-dihydro-1h-indol-2-yl-methanol, 1h-indole-2-methanol, 2,3-dihydro, 2,3-dihydro-1h-indol-2-yl methanol, 2-hydroxymethylindoline, acmc-209gye, 2-hydroxymethyl-indoline, indolin-2-ylmethan-1-ol, rs-2-hydroxymethylindoline, +-2-hydroxymethyl-indoline
C1C(NC2=CC=CC=C21)CO
149.193
149.19
2, 3-Dihydro-1H-indol-2-ylmethanol
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications