Learn More
2,3-Dimethylhydroquinone, 97%, Thermo Scientific Chemicals
105.53 € - 1207.01 €
Chemical Identifiers
CAS | 608-43-5 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00009997 |
InChI Key | BXJGUBZTZWCMEX-UHFFFAOYSA-N |
Synonym | 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone |
PubChem CID | 69100 |
IUPAC Name | 2,3-dimethylbenzene-1,4-diol |
SMILES | CC1=C(C)C(O)=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582281
|
Thermo Scientific Acros
305110050 |
5 g | Glass bottle |
105.53 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219553
|
Thermo Scientific Acros
305110250 |
25 g | Glass bottle |
359.79 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914681
|
Thermo Scientific Acros
305111000 |
100 g | Glass bottle |
1207.01 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
608-43-5 | |
138.17 | |
BXJGUBZTZWCMEX-UHFFFAOYSA-N | |
69100 | |
CC1=C(C)C(O)=CC=C1O |
C8H10O2 | |
MFCD00009997 | |
2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone | |
2,3-dimethylbenzene-1,4-diol |
Specifications
608-43-5 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD00009997 | |
Solubility in water: slightly soluble in water | |
CC1=C(C)C(O)=CC=C1O | |
138.17 | |
138.17 | |
Powder |
98.5 | |
223.0°C to 225.0°C | |
Authentic | |
C8H10O2 | |
2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone | |
BXJGUBZTZWCMEX-UHFFFAOYSA-N | |
2,3-dimethylbenzene-1,4-diol | |
69100 | |
97% | |
2, 3-Dimethylhydroquinone |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only