missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dichloroaniline, 98%, Thermo Scientific Chemicals Product Code.: 10438490

2,4-Dichloroaniline, 98%, Thermo Scientific Chemicals

Product Code. 10438490
25 g, Glass bottle
Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10438490

Brand: Thermo Scientific Acros 150390250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-00-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.02
MDL Number MFCD00007661
InChI Key KQCMTOWTPBNWDB-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin germanShow MoreShow Less
PubChem CID 11123
ChEBI CHEBI:46635
IUPAC Name 2,4-dichloroaniline
SMILES C1=CC(=C(C=C1Cl)Cl)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 59°C to 63°C
Color White to Beige
Boiling Point 242°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3NH2
Quantity 25 g
Beilstein 12,621
Solubility Information Solubility in water: insoluble
Formula Weight 162.02
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dichloroaniline, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464177 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number150392500,150390010,150390250
Lot NumberA0424579 Certificate TypeCertificate of Analysis Date21/11/2023 Catalog Number150392500,150390010,150390025,150390250
Lot NumberA0432435 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog Number150392500,150390010,150390025,150390250
Lot NumberA0403934 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150392500,150390010,150390025,150390250
Lot NumberA0402901 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150392500,150390010,150390025,150390250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dichloroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.