missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

51.10 € - 164.00 €

Chemical Identifiers

CAS 56946-83-9
Molecular Formula C4HCl3O2S2
Molecular Weight (g/mol) 251.52
MDL Number MFCD00051665
InChI Key JJKSHSHZJOWSEC-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophene-3-sulphonyl chloride, 2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonyl chloride, 2,5-dichloro, 2,5-dichlorothiophene-3-sulfonic acid chloride, pubchem7412, 2,5-dichlorothiophene-3-sulfonylchloride, acmc-1avwk, 2,5-dichloro 3-thienyl chlorosulfone, 2,5-dichloro-3-thienylsulfonyl chloride, 2,5-dichlorothien-3-yl-sulfonyl chloride
PubChem CID 2736094
IUPAC Name 2,5-dichlorothiophene-3-sulfonyl chloride
SMILES C1=C(SC(=C1S(=O)(=O)Cl)Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362526
View Documents
Thermo Scientific Alfa Aesar
L06461.03
1 g
51.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
11372526
View Documents
Thermo Scientific Alfa Aesar
L06461.06
5 g
164.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56946-83-9
251.52
JJKSHSHZJOWSEC-UHFFFAOYSA-N
2736094
C1=C(SC(=C1S(=O)(=O)Cl)Cl)Cl
C4HCl3O2S2
MFCD00051665
2,5-dichlorothiophene-3-sulphonyl chloride, 2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonyl chloride, 2,5-dichloro, 2,5-dichlorothiophene-3-sulfonic acid chloride, pubchem7412, 2,5-dichlorothiophene-3-sulfonylchloride, acmc-1avwk, 2,5-dichloro 3-thienyl chlorosulfone, 2,5-dichloro-3-thienylsulfonyl chloride, 2,5-dichlorothien-3-yl-sulfonyl chloride
2,5-dichlorothiophene-3-sulfonyl chloride

Specifications

56946-83-9
256°C to 257°C
C4HCl3O2S2
MFCD00051665
1285315
2,5-dichlorothiophene-3-sulphonyl chloride, 2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonyl chloride, 2,5-dichloro, 2,5-dichlorothiophene-3-sulfonic acid chloride, pubchem7412, 2,5-dichlorothiophene-3-sulfonylchloride, acmc-1avwk, 2,5-dichloro 3-thienyl chlorosulfone, 2,5-dichloro-3-thienylsulfonyl chloride, 2,5-dichlorothien-3-yl-sulfonyl chloride
C1=C(SC(=C1S(=O)(=O)Cl)Cl)Cl
251.52
251.54
2,5-Dichlorothiophene-3-sulfonyl chloride
1.697
>110°C (230°F)
1.597
UN3265
Moisture sensitive
JJKSHSHZJOWSEC-UHFFFAOYSA-N
2,5-dichlorothiophene-3-sulfonyl chloride
2736094
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.