missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyl-p-benzoquinone, 96%, Thermo Scientific Chemicals

33.20 € - 114.00 €

Chemical Identifiers

CAS 844-51-9
Molecular Formula C18H12O2
Molecular Weight (g/mol) 260.29
MDL Number MFCD00001600
InChI Key QYXHDJJYVDLECA-UHFFFAOYSA-N
Synonym 2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
PubChem CID 70055
IUPAC Name 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11458306
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15369.06
5 g
33.20 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11468306
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15369.14
25 g
114.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

Solubility
Soluble in hot Toluene (almost transparency).

Notes
Incompatible with Oxidizing agents, Reducing agent.
Specifications

Chemical Identifiers

844-51-9
260.29
QYXHDJJYVDLECA-UHFFFAOYSA-N
70055
O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1
C18H12O2
MFCD00001600
2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
2,5-diphenylcyclohexa-2,5-diene-1,4-dione

Specifications

844-51-9
C18H12O2
2051812
Soluble in hot Toluene (almost transparency).
O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1
260.29
260.3
2,5-Diphenyl-p-benzoquinone
215°C to 219°C
MFCD00001600
2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
QYXHDJJYVDLECA-UHFFFAOYSA-N
2,5-diphenylcyclohexa-2,5-diene-1,4-dione
70055
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

missing translation for 'einecsNumber' : 212-680-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only