Learn More
2,6-Dichloropyrazine, 98%, Thermo Scientific Chemicals
121.61 € - 1261.28 €
Chemical Identifiers
CAS | 4774-14-5 |
---|---|
Molecular Formula | C4H2Cl2N2 |
Molecular Weight (g/mol) | 148.974 |
MDL Number | MFCD00006125 |
InChI Key | LSEAAPGIZCDEEH-UHFFFAOYSA-N |
Synonym | pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine |
PubChem CID | 78504 |
IUPAC Name | 2,6-dichloropyrazine |
SMILES | C1=C(N=C(C=N1)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414310
|
Thermo Scientific Alfa Aesar
A11175.06 |
5 g |
121.61 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424310
|
Thermo Scientific Alfa Aesar
A11175.14 |
25 g |
479.39 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434310
|
Thermo Scientific Alfa Aesar
A11175.22 |
100 g |
1261.28 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Dichloropyrazine is used in the preparation of pyrazine and pyridine compounds as ATR kinase inhibitors. It is also used in the synthesis of oxacalixarenes containing nitrogen heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dichloropyrazine is used in the preparation of pyrazine and pyridine compounds as ATR kinase inhibitors. It is also used in the synthesis of oxacalixarenes containing nitrogen heterocycles.
Solubility
Soluble in ethanol and methanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids and bases.
Chemical Identifiers
4774-14-5 | |
148.974 | |
LSEAAPGIZCDEEH-UHFFFAOYSA-N | |
78504 | |
C1=C(N=C(C=N1)Cl)Cl |
C4H2Cl2N2 | |
MFCD00006125 | |
pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine | |
2,6-dichloropyrazine |
Specifications
4774-14-5 | |
>110°C (230°F) | |
MFCD00006125 | |
pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine | |
LSEAAPGIZCDEEH-UHFFFAOYSA-N | |
2,6-dichloropyrazine | |
78504 | |
99% |
52°C to 56°C | |
C4H2Cl2N2 | |
2439 | |
Soluble in ethanol and methanol. | |
C1=C(N=C(C=N1)Cl)Cl | |
148.974 | |
148.98 | |
2,6-Dichloropyrazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 225-316-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.