missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetyl-1-methylpyrrole, 98%, Thermo Scientific Chemicals
31.30 € - 438.00 €
Chemical Identifiers
CAS | 932-16-1 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.16 |
MDL Number | MFCD00003089 |
InChI Key | NZFLWVDXYUGFAV-UHFFFAOYSA-N |
Synonym | 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole |
PubChem CID | 61240 |
ChEBI | CHEBI:59982 |
IUPAC Name | 1-(1-methylpyrrol-2-yl)ethanone |
SMILES | CN1C=CC=C1C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498276
|
Thermo Scientific Alfa Aesar
A15353.06 |
5 g |
31.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408286
|
Thermo Scientific Alfa Aesar
A15353.14 |
25 g |
110.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418286
|
Thermo Scientific Alfa Aesar
A15353.22 |
100 g |
438.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
932-16-1 | |
123.16 | |
NZFLWVDXYUGFAV-UHFFFAOYSA-N | |
61240 | |
1-(1-methylpyrrol-2-yl)ethanone |
C7H9NO | |
MFCD00003089 | |
2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole | |
CHEBI:59982 | |
CN1C=CC=C1C(C)=O |
Specifications
932-16-1 | |
200°C to 202°C | |
Pleasant | |
1.542 | |
111887 | |
NZFLWVDXYUGFAV-UHFFFAOYSA-N | |
1-(1-methylpyrrol-2-yl)ethanone | |
61240 | |
123.16 | |
2-Acetyl-1-methylpyrrole |
1.04 | |
68°C (154°F) | |
C7H9NO | |
MFCD00003089 | |
2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole | |
CN1C=CC=C1C(C)=O | |
123.16 | |
CHEBI:59982 | |
98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 213-247-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only