Learn More
2-Acetylphenylboronic acid, 96%, Thermo Scientific Chemicals
51.96 € - 795.96 €
Chemical Identifiers
CAS | 308103-40-4 |
---|---|
Molecular Formula | C8H9BO3 |
Molecular Weight (g/mol) | 163.97 |
MDL Number | MFCD01321263 |
InChI Key | ZKAOVABYLXQUTI-UHFFFAOYSA-N |
Synonym | 2-acetylphenyl boronic acid, boronic acid, 2-acetylphenyl, 2'-boronoacetophenone, 1-2-boronophenyl ethan-1-one, 2-acetylbenzeneboronic acid, 2-acetylbenzenboronicacid, 1-2-dihydroxyboranyl phenyl ethan-1-one, 2-acetylphenylboronicacid, pubchem5373, acmc-1agh6 |
PubChem CID | 2734309 |
IUPAC Name | (2-acetylphenyl)boronic acid |
SMILES | CC(=O)C1=CC=CC=C1B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10487582
|
Thermo Scientific Acros
358820010 |
1 g |
51.96 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10405135
|
Thermo Scientific Acros
358820050 |
5 g |
168.84 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10780575
|
Thermo Scientific Acros
358820250 |
25 g |
795.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
308103-40-4 | |
163.97 | |
ZKAOVABYLXQUTI-UHFFFAOYSA-N | |
2734309 | |
CC(=O)C1=CC=CC=C1B(O)O |
C8H9BO3 | |
MFCD01321263 | |
2-acetylphenyl boronic acid, boronic acid, 2-acetylphenyl, 2'-boronoacetophenone, 1-2-boronophenyl ethan-1-one, 2-acetylbenzeneboronic acid, 2-acetylbenzenboronicacid, 1-2-dihydroxyboranyl phenyl ethan-1-one, 2-acetylphenylboronicacid, pubchem5373, acmc-1agh6 | |
(2-acetylphenyl)boronic acid |
Specifications
308103-40-4 | |
100.0 | |
Yellow | |
95% min. (HPLC) | |
C8H9BO3 | |
2-acetylphenyl boronic acid, boronic acid, 2-acetylphenyl, 2'-boronoacetophenone, 1-2-boronophenyl ethan-1-one, 2-acetylbenzeneboronic acid, 2-acetylbenzenboronicacid, 1-2-dihydroxyboranyl phenyl ethan-1-one, 2-acetylphenylboronicacid, pubchem5373, acmc-1agh6 | |
CC(=O)C1=CC=CC=C1B(O)O | |
163.97 | |
163.97 | |
Chunks and/or Powder |
95.0 | |
170.0°C | |
Authentic | |
Glass bottle | |
MFCD01321263 | |
ZKAOVABYLXQUTI-UHFFFAOYSA-N | |
(2-acetylphenyl)boronic acid | |
2734309 | |
96% | |
2-Acetylphenylboronic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.