Learn More
2'-Chloro-6'-fluoroacetophenone, 97%, Thermo Scientific™
36.00 € - 535.00 €
Chemical Identifiers
CAS | 87327-69-3 |
---|---|
Molecular Formula | C8H6ClFO |
Molecular Weight (g/mol) | 172.58 |
MDL Number | MFCD01631390 |
InChI Key | DNVGZKIRMBCQEQ-UHFFFAOYSA-N |
PubChem CID | 2773578 |
SMILES | CC(=O)C1=C(F)C=CC=C1Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2′-Chloro-6′-fluoroacetophenone is used in medicine & organic intermediate.
Solubility
Sparingly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
87327-69-3 | |
172.58 | |
DNVGZKIRMBCQEQ-UHFFFAOYSA-N | |
CC(=O)C1=C(F)C=CC=C1Cl |
C8H6ClFO | |
MFCD01631390 | |
2773578 |
Specifications
87327-69-3 | |
1.511 | |
5862530 | |
Sparingly soluble in water. | |
CC(=O)C1=C(F)C=CC=C1Cl | |
172.58 | |
172.59 | |
2'-Chloro-6'-fluoroacetophenone |
C8H6ClFO | |
MFCD01631390 | |
2'-chloro-6'-fluoroacetophenone, 1-2-chloro-6-fluorophenyl ethanone, 2-chloro-6-fluoroacetophenone, 1-2-chloro-6-fluorophenyl ethan-1-one, ethanone, 1-2-chloro-6-fluorophenyl, pubchem7110, 1-acetyl-2-chloro-6-fluorobenzene, 1-acetyl-6-chloro-2-fluorobenzene, 1-2-chloro-6-fluoro-phenyl-ethanone, ethanone,1-2-chloro-6-fluorophenyl | |
DNVGZKIRMBCQEQ-UHFFFAOYSA-N | |
1-(2-chloro-6-fluorophenyl)ethan-1-one | |
2773578 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.