Learn More
2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals
36.10 € - 514.00 €
Chemical Identifiers
CAS | 932-32-1 |
---|---|
Molecular Formula | C7H8ClN |
Molecular Weight (g/mol) | 141.598 |
MDL Number | MFCD00045170 |
InChI Key | WGNNILPYHCKCFF-UHFFFAOYSA-N |
Synonym | n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline |
PubChem CID | 136736 |
IUPAC Name | 2-chloro-N-methylaniline |
SMILES | CNC1=CC=CC=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11436907
|
Thermo Scientific Alfa Aesar
A14343.03 |
1 g |
36.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446907
|
Thermo Scientific Alfa Aesar
A14343.06 |
5 g |
132.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11456907
|
Thermo Scientific Alfa Aesar
A14343.14 |
25 g |
514.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson's reagent and solvent tetrahydrofuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson′s reagent and solvent tetrahydrofuran.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
932-32-1 | |
141.598 | |
WGNNILPYHCKCFF-UHFFFAOYSA-N | |
136736 | |
CNC1=CC=CC=C1Cl |
C7H8ClN | |
MFCD00045170 | |
n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline | |
2-chloro-N-methylaniline |
Specifications
932-32-1 | |
218°C | |
C7H8ClN | |
MFCD00045170 | |
2802444 | |
Not miscible or difficult to mix. | |
CNC1=CC=CC=C1Cl | |
141.598 | |
141.6 | |
2-Chloro-N-methylaniline |
1.15 | |
92°C (197°F) | |
1.58 | |
UN2810 | |
n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline | |
WGNNILPYHCKCFF-UHFFFAOYSA-N | |
2-chloro-N-methylaniline | |
136736 | |
97% |
Safety and Handling
GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only