missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 208.65 |
Percent Purity | 95% |
Chemical Name or Material | 2-(Chloromethyl)-5-(4-methylphenyl)-1, 3, 4-oxadiazole |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-(Chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole, ≥95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction