Learn More
2-Cyclohexylethanol, 99%, Thermo Scientific Chemicals
29.55 € - 433.16 €
Chemical Identifiers
CAS | 4442-79-9 |
---|---|
Molecular Formula | C8H16O |
Molecular Weight (g/mol) | 128.215 |
MDL Number | MFCD00001525 |
InChI Key | QJQZRLXDLORINA-UHFFFAOYSA-N |
Synonym | cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol |
PubChem CID | 20508 |
IUPAC Name | 2-cyclohexylethanol |
SMILES | C1CCC(CC1)CCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11465676
|
Thermo Scientific Alfa Aesar
B20782.18 |
50 g |
29.55 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475676
|
Thermo Scientific Alfa Aesar
B20782.30 |
250 g |
119.60 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455676
|
Thermo Scientific Alfa Aesar
B20782.0B |
1000 g |
433.16 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a flavoring agent.
Solubility
Soluble in alcohol and dipropylene glycol. Soluble in water (1750 mg/L at 25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
4442-79-9 | |
128.215 | |
QJQZRLXDLORINA-UHFFFAOYSA-N | |
20508 | |
C1CCC(CC1)CCO |
C8H16O | |
MFCD00001525 | |
cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol | |
2-cyclohexylethanol |
Specifications
4442-79-9 | |
0.92 | |
86°C (186°F) | |
1.466 | |
1848152 | |
Soluble in alcohol and dipropylene glycol. Soluble in water (1750mg/L at 25°C). | |
C1CCC(CC1)CCO | |
128.215 | |
128.22 | |
2-Cyclohexylethanol |
-20°C | |
206°C to 207°C | |
C8H16O | |
MFCD00001525 | |
cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol | |
QJQZRLXDLORINA-UHFFFAOYSA-N | |
2-cyclohexylethanol | |
20508 | |
99% |
Safety and Handling
GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'einecsNumber' : 224-672-1
missing translation for 'rtecsNumber' : KK3528000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only