Learn More
2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals
71.90 € - 255.00 €
Chemical Identifiers
CAS | 321-60-8 |
---|---|
Molecular Formula | C12H9F |
Molecular Weight (g/mol) | 172.20 |
MDL Number | MFCD00000317 |
InChI Key | KLECYOQFQXJYBC-UHFFFAOYSA-N |
Synonym | 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene |
PubChem CID | 67579 |
SMILES | FC1=CC=CC=C1C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404913
|
Thermo Scientific Alfa Aesar
A12227.06 |
5 g |
71.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319619
|
Thermo Scientific Alfa Aesar
A12227.09 |
10 g |
128.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414913
|
Thermo Scientific Alfa Aesar
A12227.14 |
25 g |
255.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
Solubility
Soluble in alcohol, ether. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Chemical Identifiers
321-60-8 | |
172.20 | |
KLECYOQFQXJYBC-UHFFFAOYSA-N | |
67579 |
C12H9F | |
MFCD00000317 | |
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
FC1=CC=CC=C1C1=CC=CC=C1 |
Specifications
321-60-8 | |
1.245 | |
C12H9F | |
2043175 | |
Soluble in alcohol,ether. Insoluble in water. | |
FC1=CC=CC=C1C1=CC=CC=C1 | |
172.20 | |
172.2 | |
2-Fluorobiphenyl |
72°C to 76°C | |
248°C | |
MFCD00000317 | |
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
KLECYOQFQXJYBC-UHFFFAOYSA-N | |
1-fluoro-2-phenylbenzene | |
67579 | |
98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 206-290-7
missing translation for 'rtecsNumber' : DV5291000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only