missing translation for 'onlineSavingsMsg'
Learn More
2-Imidazolidinone hemihydrate, 98+%, Thermo Scientific Chemicals Product Code.: 11423248

2-Imidazolidinone hemihydrate, 98+%, Thermo Scientific Chemicals

Product Code. 11423248
1000 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11423248

Brand: Thermo Scientific Alfa Aesar A17666.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121325-67-5
Molecular Formula C6H14N4O3
Molecular Weight (g/mol) 190.20
MDL Number MFCD02662355
InChI Key KPRJGGOOWATRNT-UHFFFAOYSA-N
Synonym imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrateShow MoreShow Less
PubChem CID 2723651
IUPAC Name imidazolidin-2-one;hydrate
SMILES O.O=C1NCCN1.O=C1NCCN1

Specifications

Melting Point 55°C
Quantity 1000 g
Solubility Information Soluble in water.
Formula Weight 95.10 (86.09 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 2-Imidazolidinone hemihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10210117 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberA17666.30,A17666.0B
Lot Number10212949 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA17666.30,A17666.0B
Lot Number10187478 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17666.30,A17666.0B
Lot Number10099647 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17666.30,A17666.0B
Lot Number10103397 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17666.30,A17666.0B
compliance-icons compliance-icons
Product Identifier
  • 2-Imidazolidinone hemihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Imidazolidinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Imidazolidinone hemihydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.