missing translation for 'onlineSavingsMsg'
Learn More
2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals Product Code.: 10051260

2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals

Product Code. 10051260
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10051260

Brand: Thermo Scientific Acros 125561000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-53-7
Molecular Formula C3H5NS2
Molecular Weight (g/mol) 119.21
MDL Number MFCD00126013
InChI Key WGJCBBASTRWVJL-UHFFFAOYSA-N
Synonym 2-mercaptothiazoline, 2-thiazolidinethione, 2-thiazoline-2-thiol, thiazolidine-2-thione, metabasal, thyroidan, 2-mercapto-2-thiazoline, mercaptothiazoline, 2-thiothiazolidone, 2-thiazolidenethioneShow MoreShow Less
PubChem CID 2723699
IUPAC Name 1,3-thiazolidine-2-thione
SMILES C1CSC(=S)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 103°C to 106°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 27, 140
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alkaline water and acetic acid,soluble in ethanol,benzene and chcl3,slightly soluble in ether,insoluble in cs2
Formula Weight 119.21
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Mercaptothiazoline, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0413812 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number125565000,125561000
Lot NumberA0429338 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog Number125565000,12556015,12556023,125561000,125560050
Lot NumberA0403644 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number125565000,12556015,12556023,125561000,125560050
Lot NumberA0413812 Certificate TypeCertificate of Analysis Date27/05/2022 Catalog Number125565000,12556015,12556023,125561000,125560050
Lot NumberA0393978 Certificate TypeCertificate of Analysis Date04/09/2020 Catalog Number125565000,12556015,12556023,125561000,125560050
compliance-icons
Product Identifier
  • 2-Mercaptothiazoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.