Learn More
Rhodanine, 98+%, Thermo Scientific Chemicals
Description
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.
Solubility
Soluble in water (2.25 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 166°C to 171°C |
Density | 0.868 |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 110701 |
Merck Index | 14,8184 |
Solubility Information | Soluble in water (2.25g/L at 20°C). |
Formula Weight | 133.19 |
Percent Purity | ≥98% |
Chemical Name or Material | Rhodanine |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
GBFA019684 | Certificate of Analysis | 23/11/2021 | A18293.14,A18293.36,A18293.22 |
J08T046 | Certificate of Analysis | 23/11/2021 | A18293.14,A18293.36,A18293.22 |
10141697 | Certificate of Analysis | 23/11/2021 | A18293.14,A18293.36,A18293.22 |
10156461 | Certificate of Analysis | 23/11/2021 | A18293.14,A18293.36,A18293.22 |
10177050 | Certificate of Analysis | 23/11/2021 | A18293.14,A18293.36,A18293.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.