missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzeneboronic acid 1,3-propanediol ester, 95%, Thermo Scientific™

30.00 € - 507.00 €

Chemical Identifiers

CAS 163517-56-4
Molecular Formula C10H13BO2
Molecular Weight (g/mol) 176.022
MDL Number MFCD06659935
InChI Key UIDYFYHQHHMEKA-UHFFFAOYSA-N
Synonym 2-2-methylphenyl-1,3,2-dioxaborinane, 2-methylbenzeneboronic acid, propanediol cyclic ester, 2-methylphenylboronic acid propanediol cyclic ester, 2-o-tolyl-1,3,2-dioxaborinane, 2-2-methylphenyl 1,3,2-dioxaborinane, 1,3,2-dioxaborinane,2-2-methylphenyl, 2-methylbenzeneboronic acid 1,3-propanediol ester, 2-methylphenylboronic acid,propanediol cyclic ester, 2-methylphenylboronic acid, propanediol cyclic ester
PubChem CID 23005365
IUPAC Name 2-(2-methylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCCCO1)C2=CC=CC=C2C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15480056
View Documents
Thermo Scientific Alfa Aesar
H52720.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15490056
View Documents
Thermo Scientific Alfa Aesar
H52720.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15400066
View Documents
Thermo Scientific Alfa Aesar
H52720.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

163517-56-4
176.022
UIDYFYHQHHMEKA-UHFFFAOYSA-N
23005365
B1(OCCCO1)C2=CC=CC=C2C
C10H13BO2
MFCD06659935
2-2-methylphenyl-1,3,2-dioxaborinane, 2-methylbenzeneboronic acid, propanediol cyclic ester, 2-methylphenylboronic acid propanediol cyclic ester, 2-o-tolyl-1,3,2-dioxaborinane, 2-2-methylphenyl 1,3,2-dioxaborinane, 1,3,2-dioxaborinane,2-2-methylphenyl, 2-methylbenzeneboronic acid 1,3-propanediol ester, 2-methylphenylboronic acid,propanediol cyclic ester, 2-methylphenylboronic acid, propanediol cyclic ester
2-(2-methylphenyl)-1,3,2-dioxaborinane

Specifications

163517-56-4
MFCD06659935
UIDYFYHQHHMEKA-UHFFFAOYSA-N
2-(2-methylphenyl)-1,3,2-dioxaborinane
23005365
95%
C10H13BO2
2-2-methylphenyl-1,3,2-dioxaborinane, 2-methylbenzeneboronic acid, propanediol cyclic ester, 2-methylphenylboronic acid propanediol cyclic ester, 2-o-tolyl-1,3,2-dioxaborinane, 2-2-methylphenyl 1,3,2-dioxaborinane, 1,3,2-dioxaborinane,2-2-methylphenyl, 2-methylbenzeneboronic acid 1,3-propanediol ester, 2-methylphenylboronic acid,propanediol cyclic ester, 2-methylphenylboronic acid, propanediol cyclic ester
B1(OCCCO1)C2=CC=CC=C2C
176.022
176.02
2-Methylbenzeneboronic acid 1,3-propanediol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications