Learn More
2-Methylheptane, 99%, Thermo Scientific Chemicals
55.90 € - 740.00 €
Chemical Identifiers
CAS | 592-27-8 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
MDL Number | MFCD00009518 |
InChI Key | JVSWJIKNEAIKJW-UHFFFAOYSA-N |
Synonym | heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3 |
PubChem CID | 11594 |
IUPAC Name | 2-methylheptane |
SMILES | CCCCCC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356649
|
Thermo Scientific Alfa Aesar
H26220.03 |
1 g |
55.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366649
|
Thermo Scientific Alfa Aesar
H26220.06 |
5 g |
200.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376649
|
Thermo Scientific Alfa Aesar
H26220.14 |
25 g |
740.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Chemical Identifiers
592-27-8 | |
114.232 | |
JVSWJIKNEAIKJW-UHFFFAOYSA-N | |
11594 | |
CCCCCC(C)C |
C8H18 | |
MFCD00009518 | |
heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3 | |
2-methylheptane |
Specifications
592-27-8 | |
116°C | |
C8H18 | |
1.395 | |
UN1262 | |
heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3 | |
JVSWJIKNEAIKJW-UHFFFAOYSA-N | |
2-methylheptane | |
11594 | |
99% |
0.698 | |
4°C (40°F) | |
(CH3)2CHCH2CH2CH2CH2CH3 | |
MFCD00009518 | |
1696862 | |
Soluble in alcohol. Insoluble in water. | |
CCCCCC(C)C | |
114.232 | |
114.23 | |
2-Methylheptane |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
missing translation for 'einecsNumber' : 209-747-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only