missing translation for 'onlineSavingsMsg'
Learn More
2-(Methylthio)aniline, 98%, Thermo Scientific Chemicals Product Code.: 11475093

2-(Methylthio)aniline, 98%, Thermo Scientific Chemicals

Product Code. 11475093
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11475093

Brand: Thermo Scientific Alfa Aesar A12762.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (less).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2987-53-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007708
InChI Key WBRPQQSADOCKCH-UHFFFAOYSA-N
Synonym 2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio anilineShow MoreShow Less
PubChem CID 76337
IUPAC Name 2-methylsulfanylaniline
SMILES CSC1=CC=CC=C1N

Specifications

Density 1.111
Boiling Point 233°C to 234°C
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 5 g
UN Number UN2810
Beilstein 386211
Solubility Information Soluble in water (less).
Formula Weight 139.23
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)aniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10205910 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberA12762.22,A12762.14,A12762.06
Lot Number10169524 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12762.22,A12762.14,A12762.06
Lot Number10205910 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12762.22,A12762.14,A12762.06
Lot Number10201058 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12762.22,A12762.14,A12762.06
Lot Number5003R04M Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12762.22,A12762.14,A12762.06
compliance-icons
Product Identifier
  • 2-(Methylthio)aniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : 2-(Methylthio)aniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Methylthio)aniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.