missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ 2 mL Class AS Bang Straight Stopcock Micro Burettes

With Bang pattern Schellbach stripes for enhanced legibility.

Brand:  Hirschmann™ 3300140

Product Code. 13108009

  • 326.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class AS
  • Manufactured from DURAN™ borosilicate glass for resistance to chemicals and thermal shock
  • Blue graduation marks
  • Certified conformity, with dated batch identification
  • Straight stopcock with standard ground glass key
  • Stopcock in filling tube with glass key
  • Main point ring graduation
  • Calibrated for EX
  • Acc. to DIN EN ISO 385
Specifications

Specifications

2 mL
0.01 mL
Blue
Glass
Microburet
DIN EN ISO 385
Borosilicate Glass
Bang, Schellbach Stripes
Straight
±0.01 mL
Class AS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ 2 mL Class AS Bang Straight Stopcock Micro Burettes > Capacity: 2 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.