missing translation for 'onlineSavingsMsg'
Learn More

2’-O,4’-C-Methyleneuridine, MedChemExpress

MedChemExpress 2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.

209.25 € - 348.30 €

Specifications

Chemical Name or Material 2’-O,4’-C-Methyleneuridine
CAS 200435-92-3
Color White
Purity Grade Notes Research
Molecular Formula C10H12N2O6
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16430817
Encompass_Preferred
View Documents
Medchem Express
HY-111639
50 mg
348.30 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16414508
Encompass_Preferred
View Documents
Medchem Express
HY-111639
10 mg
209.25 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.
Specifications

Specifications

2’-O,4’-C-Methyleneuridine
White
C10H12N2O6
OC[C@@]12O[C@@H](N3C(NC(C=C3)=O)=O)[C@H](OC2)[C@@H]1O
256.21
Research
Neuroscience-Neuromodulation
200435-92-3
Research
DMSO : 125 mg/mL (487.88 mM; Need ultrasonic)
256.21
99.49%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.