missing translation for 'onlineSavingsMsg'
Learn More

(2-Phenyl-1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

118.69 € - 537.41 €

Chemical Identifiers

CAS 165736-03-8
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD02682055
InChI Key PMFCMLGAPHZRTE-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
PubChem CID 736526
IUPAC Name (2-phenyl-1,3-thiazol-4-yl)methanamine
SMILES C1=CC=C(C=C1)C2=NC(=CS2)CN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10079231
View Documents
Thermo Scientific Maybridge
CC18313DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10041124
View Documents
Thermo Scientific Maybridge
CC18313DE
5 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10356304
View Documents
Thermo Scientific Maybridge
CC18313EA
10 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

165736-03-8
190.264
PMFCMLGAPHZRTE-UHFFFAOYSA-N
736526
C1=CC=C(C=C1)C2=NC(=CS2)CN
C10H10N2S
MFCD02682055
2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
(2-phenyl-1,3-thiazol-4-yl)methanamine

Specifications

165736-03-8
100.0
C10H10N2S
2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
C1=CC=C(C=C1)C2=NC(=CS2)CN
190.264
190.27
(2-Phenyl-1, 3-thiazol-4-yl)methylamine
96.5
97%
MFCD02682055
PMFCMLGAPHZRTE-UHFFFAOYSA-N
(2-phenyl-1,3-thiazol-4-yl)methanamine
736526
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.