Learn More
Ismatec™ 2-Stop Tubing, Tygon HC F-4040-A
Special tubing for hydrocarbons, petroleum products, and distillates.
Brand: Ismatec™ SC0166
This item is not returnable.
View return policy
Description
- Specially formulated to transport hydrocarbons, petroleum products and distillates.
- Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.
- Low gas permeability.
Specifications
Yellow-Blue | |
Tygon HC | |
Pumps that incorporate cassettes into the pumphead design. Hydrocarbons, petroleum products, and distillates. | |
12/Pk | |
I.D. 1.52 mm | |
74°C |
1.52 mm | |
400 mm | |
−37°C | |
Acids: Limited, Alkaline solutions: Not rec., Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited | |
Amber |