missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trichloroacetyl)pyrrole, 98%, Thermo Scientific™
50.93 € - 189.00 €
Chemical Identifiers
CAS | 35302-72-8 |
---|---|
Molecular Formula | C6H4Cl3NO |
Molecular Weight (g/mol) | 212.46 |
MDL Number | MFCD00128757 |
InChI Key | BBFDGMDENAEMKF-UHFFFAOYSA-N |
Synonym | 2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole |
PubChem CID | 321487 |
IUPAC Name | 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone |
SMILES | C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl |
Description
Chemical Identifiers
35302-72-8 | |
212.46 | |
BBFDGMDENAEMKF-UHFFFAOYSA-N | |
321487 | |
C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl |
C6H4Cl3NO | |
MFCD00128757 | |
2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole | |
2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone |
Specifications
35302-72-8 | |
100.0 | |
Green | |
Authentic | |
C6H4Cl3NO | |
2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole | |
C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl | |
212.46 | |
212.46 | |
Crystalline Powder |
97.5 | |
72.0°C to 74.0°C | |
33.07 to 34.76% C, 6.43 to 7.75% N | |
98% | |
MFCD00128757 | |
BBFDGMDENAEMKF-UHFFFAOYSA-N | |
2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone | |
321487 | |
98% | |
2-(Trichloroacetyl)pyrrole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-(Trichloroacetyl)pyrrole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only