Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand:  RETSCH 60.123.012500
 View more versions of this product
Product Code. 10075001
  • 173.00 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
25 mm
200 mm
Stainless Steel
12.5 mm
ASTM E 11
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel