Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand:  RETSCH
 View more versions of this product
Product Code. 10638571
  • 174.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
25 mm
200 mm
Stainless Steel
75 mm
ASTM E 11
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel