Learn More
RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ASTM Certificate
Brand:  RETSCH 60.150.106000
 View more versions of this product
Product Code. 10476621
  • 174.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
50.8mm
203.2 mm
Stainless Steel
106 mm
ASTM E 11
Shaker AS 200 Tap
For separation, fractioning, particle size determination
Stainless Steel