missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 203mm Diameter Stainless Steel Test Sieve with Electroformed Sheet

Brand:  RETSCH 60.142.000016

 View more versions of this product

Product Code. 11974412

  • 3405.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

1Each
Test Sieve
Stainless Steel
25mm
203 mm
Stainless Steel
16 μm
ISO 3310/3
Air Jet Sieving Machine AS 200 jet
For separation, fractioning, particle size determination
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.