missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 20mm Crimp Cap

Aluminum crimp closures provide a secure leak-resistant seal.

Brand:  Fisherbrand™ 20 01 0211

 View more versions of this product

Product Code. 15440130

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • 20mm
  • aluminum Cap
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Center Tear-off
20 mm
100
ND20
Red Lacquered
Aluminum
Crimp Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.