missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 20X Bolt™ MES SDS Running Buffer

Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats

74.00 € - 376.00 €

Specifications

Concentration 20X
For Use With (Equipment) Bolt
Product Line Bolt
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13266499
View Documents
Invitrogen™
B0002
500 mL
74.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
13246499
View Documents
Invitrogen™
B000202
5 L
376.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bolt™ buffers and sample reducing agents are optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats. Note: Bolt™ gel run times may vary depending upon the type of running buffer used.

About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.
Specifications

Specifications

20X
Bolt
Bolt
Videos
Safety and Handling

Safety and Handling

Recommended Storage : MES SDS Running Buffer (20X).

Store at room temperature.

Shelf-life is 6 months, when stored properly.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.