missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Polycarbonate Protective Safety Glasses, 2720 Series
Provides reliable protection against specified levels of hazardous UV radiation. The 3M Polycarbonate Protective Safety safety glasses are lightweight glasses with a sporty, modern design.
6.03 € - 157.80 €
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
---|---|
Product Type | Safety glasses |
For Use With (Application) | Construction, Metal Fabrication, forestry |
Frame Color | Blue |
No. per Pack | 1 Ea. |
Product Code | Brand | Lens Tint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Tint | Price | Quantity & Availability | ||||||
10667435
|
3M™
7000032452 |
Amber | N/A | N/A | N/A | |||||
10126394
|
3M™
7000032451 |
Gray | N/A | N/A | N/A | |||||
10024274
|
3M™
7000032450 |
Clear | N/A | N/A | N/A | |||||
16391696
|
3M™
7000032450 |
Clear | N/A | N/A | N/A | |||||
11786003
|
3M™
K2720 |
Clear | N/A | N/A | N/A | |||||
Description
Extremely lightweight and comfortable with a sporty, modern design.- Optical Class 1 for high optical clarity
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Anti-scratch coating for increased durability and improved vision
- Anti-fog coating to help reduce lens fogging, particularly during physical work or when wearing half masks
- Ideal for use in many environments, including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking
- Combines well with 3M™ Aura™ 9300+ Series Disposable Respirators
Specifications
Anti-fog, Scratch-resistant | |
Construction, Metal Fabrication, forestry | |
1 Ea. |
Safety glasses | |
Blue | |
Polycarbonate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Polycarbonate Protective Safety Glasses, 2720 Series