Learn More
3M™ Polycarbonate Protective Safety Glasses, 2720 Series
Provides reliable protection against specified levels of hazardous UV radiation. The 3M Polycarbonate Protective Safety safety glasses are lightweight glasses with a sporty, modern design.
Brand: 3M™ 7000032450
Description
Extremely lightweight and comfortable with a sporty, modern design.- Optical Class 1 for high optical clarity
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Anti-scratch coating for increased durability and improved vision
- Anti-fog coating to help reduce lens fogging, particularly during physical work or when wearing half masks
- Ideal for use in many environments, including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking
- Combines well with 3M™ Aura™ 9300+ Series Disposable Respirators
Specifications
Anti-fog, Scratch-resistant | |
Safety glasses | |
Blue | |
Polycarbonate |
Clear | |
Construction, Metal Fabrication, forestry | |
1 Ea. |