Learn More
2M Lithium Chloride Electrolyte, Electrode Filling Solution, Paragon Scientific™
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.
Brand:  Paragon Scientific LCL-30-3
 View more versions of this product
Product Code. 16060200
  • 154.00 € / Pack of 3
Explore more special offers
Description
Description
  • A Lithium Chloride Electrolyte Analytical Reagent manufactured to ASTM D 664 / IP 177.
  • Provided with a certificate which includes an estimation of uncertainty of manufacture.
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
  • Convenient 30mL bottle size for direct filling into the electrode.
  • Multi pack option available
  • Cost effective and time saving
  • Certificate of Manufacture & MSDS included
  • 6 Month shelf life
Specifications
Specifications
3 x 30 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
2M Lithium Chloride Electrolyte, Electrode Filling Solution, Paragon Scientific™ > Quantity: 3 x 30mL