missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2M Lithium Chloride Electrolyte, Electrode Filling Solution, Paragon Scientific™
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.
154.00 € - 317.00 €
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16060200
|
Paragon Scientific
LCL-30-3 |
3 x 30 mL |
154.00 €
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16070200
|
Paragon Scientific
LCL-30-5 |
5 x 30 mL |
202.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16050200
|
Paragon Scientific
LCL-30-10 |
10 x 30 mL |
317.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
- A Lithium Chloride Electrolyte Analytical Reagent manufactured to ASTM D 664 / IP 177.
- Provided with a certificate which includes an estimation of uncertainty of manufacture.
- Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
- Convenient 30mL bottle size for direct filling into the electrode.
- Multi pack option available
- Cost effective and time saving
- Certificate of Manufacture & MSDS included
- 6 Month shelf life
Seguridad y manipulación
missing translation for 'shelfLife' : 6 Months
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
Otros peligros
- 2M Lithium Chloride Electrolyte, Electrode Filling Solution
Palabra de advertencia
- Danger
Clasificación
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Indicaciones de peligro
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Otros peligros
- MIXTURE LIST-Contains : ethanol, lithium chloride
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto